missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NME1/NME2 Monoclonal Antibody (642825)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561615

Product Code. 30533966

  • £357.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

NME1/NME2
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
P15531, P15532, P19804, P22392, Q01768, Q05982
NME1, Nme2
E. coli-derived recombinant human NM23-H1/H2, Ala2-Glu152.
100 μg
Primary
Human, Mouse, Rat
Antibody
IgG2b
Western Blot
642825
Unconjugated
NME1
AL024257; AWD; C-myc purine-binding transcription factor PUF; c-myc transcription factor; epididymis secretory sperm binding protein Li 155an; expressed in non-metastatic cells 1 protein; expressed in non-metastatic cells 1 protein (NM23A) (nucleoside diphosphate kinase); expressed in non-metastatic cells 1, protein; expressed in non-metastatic cells 1, protein (NM23A) (nucleoside diphosphate kinase); expressed in non-metastatic cells 2; expressed in non-metastatic cells 2 protein; GAAD; Granzyme A-activated DNase; HEL-S-155an; histidine protein kinase NDKB; Metastasis inhibition factor nm23; NB; NBS; NDK A; NDK B; NDKA; NDKB; NDP kinase A; NDP kinase alpha; NDP kinase B; NDP kinase beta; NDPKA; NDPK-A; NDPKB; NDPK-B; NM23; nm23-2; NM23A; NM23B; NM23-H1; nm23-H2; NM23-M1; nm23-M2; NME/NM23 nucleoside diphosphate kinase 1; NME/NM23 nucleoside diphosphate kinase 2; NME1; NME2; non-metastatic cells 1, protein (NM23A) expressed in; non-metastatic cells 2, protein (NM23) expressed in; non-metastatic cells 2, protein (NM23B) expressed in; nucleoside diphosphate kinase A; Nucleoside diphosphate kinase B; nucleoside-diphosphate kinase 1; nucleoside-diphosphate kinase 2; nucleotide diphosphate kinase; nucleotide diphosphate kinase B; nucloside diphosphate kinase; P18; p18-12d; PUF; Tumor metastatic process-associated protein
Mouse
Protein A/G
RUO
18102, 18103, 191575, 4830, 4831, 83782
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.