missing translation for 'onlineSavingsMsg'
Learn More

NMDI14, MedChemExpress

Product Code. 16450747
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16450747

Brand: Medchem Express HY111374

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress NMDI14 is a nonsense mediated RNA decay (NMD) inhibitor.

NMDI14 is a nonsense mediated RNA decay (NMD) inhibitor.

Specifications

Chemical Name or Material NMDI14
Physical Form Solid
CAS 307519-88-6
Color Yellow
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C21H25N3O4S
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 25 mg/mL (60.17 mM)
SMILES O=C(C1=C(NC(CC2NC3=C(C=C(C)C(C)=C3)NC2=O)=O)SC(C)=C1C)OCC
Molecular Weight (g/mol) 415.51
Formula Weight 415.51
Percent Purity 95.1%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.