missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMDARA1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-89677
This item is not returnable.
View return policy
Description
NMDARA1 Polyclonal specifically detects NMDARA1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| NMDARA1 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| glutamate [NMDA] receptor-associated protein 1, glutamate receptor, ionotropic, N-methyl D-aspartate-associated protein 1(glutamate binding), glutamate receptor, NMDA subtype, glutamate-binding subunit, HNRGW, LFG1, NMDA receptor glutamate-binding subunit, NMDARA1MGC99687, Putative MAPK-activating protein PM02, TMBIM3, transmembrane BAX inhibitor motif containing 3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 2907 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| GRINA | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:PYAQPPYPGAPYPQPPFQPSPYGQPGYPHGPSPYPQGGYPQGPY | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction