missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMDAR3B Antibody (2A8E11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£173.00 - £453.00
Specifications
| Antigen | NMDAR3B |
|---|---|
| Clone | 2A8E11 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 |
| Applications | Western Blot, Flow Cytometry, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18213851
|
Novus Biologicals
NBP2-61747 |
0.1 mL |
£453.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18669868
|
Novus Biologicals
NBP2-61747-0.025ml |
0.025 mL |
£173.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NMDAR3B Monoclonal specifically detects NMDAR3B in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA.Specifications
| NMDAR3B | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA | |
| Unconjugated | |
| Mouse | |
| GRIN3B | |
| IgG2a | |
| Protein G purified |
| 2A8E11 | |
| Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000 | |
| Monoclonal | |
| Purified | |
| 116444 | |
| Purified recombinant fragment of human GRIN3B (AA: 135-276) expressed in E. Coli. | |
| Primary | |
| 113 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title