missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMDAR2B, p Ser1480 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-61103
This item is not returnable.
View return policy
Description
NMDAR2B Polyclonal specifically detects NMDAR2B in Mouse, Rat samples. It is validated for Western Blot.
Specifications
| NMDAR2B (p Ser1480) | |
| Polyclonal | |
| Western Blot 1:1000 | |
| Q00960 | |
| GRIN2B | |
| Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser1480 conjugated to KLH. Accession # Q00960 | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Specific for ∼180k NMDAR NR2B subunit protein phosphorylated at Ser1480. Immunolabeling of the NMDA NR2B subunit band is blocked by the phosphopeptide used as the antigen but not by the corresponding dephosphopeptide. Immunolabeling is also blocked by lambda-phosphatase treatment. | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| 10mM HEPES (pH 7.5), 0.15M NaCl, 0.1 mg/ml BSA and 50% Glycerol with No Preservative | |
| glutamate [NMDA] receptor subunit epsilon-2, glutamate receptor subunit epsilon-2, glutamate receptor, ionotropic, N-methyl D-aspartate 2B, hNR3, MGC142180, NMDAR2BMGC142178, N-methyl D-aspartate receptor subtype 2B, N-methyl-D-aspartate receptor subunit 3, NR2B, NR3 | |
| Rabbit | |
| 180 kDa | |
| 0.1 mL | |
| Neurodegeneration, Neuronal Cell Markers, Neuroscience, Phospho Specific | |
| 2904 | |
| Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction