missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMDAR2B Antibody (17), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43543
This item is not returnable.
View return policy
Description
NMDAR2B Monoclonal antibody specifically detects NMDAR2B in Rat samples. It is validated for X-Ray Crystallography
Specifications
| NMDAR2B | |
| Monoclonal | |
| 1 mg/mL | |
| X-Ray Crystallography | |
| glutamate [NMDA] receptor subunit epsilon-2, glutamate receptor subunit epsilon-2, glutamate receptor, ionotropic, N-methyl D-aspartate 2B, hNR3, MGC142180, NMDAR2BMGC142178, N-methyl D-aspartate receptor subtype 2B, N-methyl-D-aspartate receptor subunit 3, NR2B, NR3 | |
| Monoclonal antibodies that bind rat GluN2B ATD were obtained by immunizing mice with the purified intact GluN1-GluN2B NMDA receptors. IgGs were purified from hybridoma cell culture supernatant by Protein-A Sepharose. Fab fragments have been prepared by papain proteolysis followed by re-chromatographing onto Protein-A Sepharose to remove Fc. (Uniprot# Q00960) | |
| 0.2 mg | |
| Autophagy, Neurodegeneration, Neuroendocrinology, Neuronal Cell Markers, Neuroscience, Phospho Specific | |
| 2904 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| X-Ray Crystallography | |
| 17 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction