missing translation for 'onlineSavingsMsg'
Learn More

NMDAR2A Antibody (T7), Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18424575 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18424575 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18424575 Supplier Novus Biologicals Supplier No. NBP229863AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NMDAR2A Monoclonal antibody specifically detects NMDAR2A in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen NMDAR2A
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone T7
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias GluN2A, glutamate receptor, ionotropic, N-methyl D-aspartate 2A, hNR2A, NMDA receptor subtype 2A, N-methyl D-aspartate receptor subtype 2A, N-methyl-D-aspartate receptor subunit 2A, subunit epsilon-1
Host Species Mouse
Immunogen Amino acid region 1099-1213 from human NMDAR2A produced as a GST fusion protein.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroendocrinology, Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision
Primary or Secondary Primary
Gene ID (Entrez) 2903
Target Species Human, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.