missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NMDAR2A Antibody (S327-95), Novus Biologicals™
Mouse Monoclonal Antibody
£176.00
Specifications
| Antigen | NMDAR2A |
|---|---|
| Clone | S327-95 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence |
Description
NMDAR2A Monoclonal specifically detects NMDAR2A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| NMDAR2A | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| GluN2A, glutamate receptor, ionotropic, N-methyl D-aspartate 2A, hNR2A, NMDA receptor subtype 2A, N-methyl D-aspartate receptor subtype 2A, N-methyl-D-aspartate receptor subunit 2A, subunit epsilon-1 | |
| GRIN2A | |
| IgG2a | |
| Protein G purified | |
| Detects approx 170kDa. Does not react with NR2B. |
| S327-95 | |
| Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Q00959 | |
| 2903 | |
| Fusion protein amino acids 75-325 (extracellular N-terminus) of rat GluN2A/NR2A | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title