missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NMDA

Product Code. p-4785239
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
This item is not returnable. View return policy

Product Code. 18085632

Brand: Tocris Bioscience™ 0114/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective NMDA agonist

Prototypic NMDA receptor agonist.

Caged version also available (Cat. No 2224).

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6384-92-5
Molecular Formula C5H9NO4

Specifications

Quantity 50 mg
Formula Weight 147.13
Purity >99%
Molecular Weight (g/mol) 147.13
Chemical Name or Material NMDA
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.