missing translation for 'onlineSavingsMsg'
Learn More

NMDA, MedChemExpress

Product Code. 16427366
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
Unit Size:
100mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16427366 100 mg 100mg
16403427 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16427366 Supplier Medchem Express Supplier No. HY17551
 Restricted Item
Estimated Shipment: 26-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress NMDA is a specific agonist for NMDA receptor mimicking the action of glutamate, the neurotransmitter which normally acts at that receptor.

NMDA is a specific agonist for NMDA receptor mimicking the action of glutamate, the neurotransmitter which normally acts at that receptor.

Specifications

Chemical Name or Material NMDA
Physical Form Powder
CAS 6384-92-5
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C5H9NO4
For Use With (Application) Neuroscience-Neurodegeneration
Synonym N-Methyl-D-aspartic acid
Solubility Information H2O : 33.33 mg/mL (226.53 mM; Need ultrasonic) ∣DMSO : 10 mg/mL (67.97 mM; Need ultrasonic)
SMILES O=C(O)C[C@H](C(O)=O)NC
Molecular Weight (g/mol) 147.13
Formula Weight 147.13
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.