missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NMBR Monoclonal Antibody (466505)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA524187
This item is not returnable.
View return policy
Description
This antibody stains human Neuromedin B R/NMBR transfectants but not irrelevant transfectants. Reconstitute at 0.5 mg/mL in sterile PBS.
NMBR, also known as neuromedin B receptor, a Bombesin Receptor, activates phospholipase and protein kinase C and enhances calcium mobilization. It binds neuromedin B, a potent mitogen and growth factor for normal and neoplastic lung and for gastrointestinal epithelial tissues. Neuromedin B receptor expression has been reported in brain, particularly the olfactory bulb and thalamus, and in colon, liver, placenta, testis, spinal cord, and gastrointestinal tract. In addition, it has been shown to be present in cancer of the lung, ovary, and prostate. ESTs have been isolated from B-cell/lung/testis and brain libraries.
Specifications
| NMBR | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| P28336 | |
| NMBR | |
| NS0 mouse myeloma cell line transfected with human NMBR Met1-Met390. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry | |
| 466505 | |
| Unconjugated | |
| NMBR | |
| BB1; BB182387; BB1R; bombesin receptor 1; epididymis secretory sperm binding protein Li 185a; epididymis tissue protein Li 185a; neuromedin B receptor; neuromedin-B receptor; Neuromedin-B-preferring bombesin receptor; NMBR; NMB-R | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 4829 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction