missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NM23-H2/NME2 Rabbit anti-Mouse, Rat, Clone: 7L3A2, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | NM23-H2/NME2 |
|---|---|
| Clone | 7L3A2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18391747
|
Novus Biologicals
NBP3-16606-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18319942
|
Novus Biologicals
NBP3-16606-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NM23-H2/NME2 Monoclonal antibody specifically detects NM23-H2/NME2 in Mouse, Rat samples. It is validated for Western BlotSpecifications
| NM23-H2/NME2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 4831 | |
| IgG | |
| Affinity purified |
| 7L3A2 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| EC 2.7.13.3, Histidine protein kinase NDKB, MGC2212, NDK B, NDKB, NDPKB, NDPK-B, nm23-H2, non-metastatic cells 2, protein (NM23B) expressed in, nucleoside diphosphate kinase B, protein (NM23) expressed in | |
| A synthetic peptide corresponding to a sequence within amino acids 50-152 of human NM23-H2/NME2 (P22392). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title