missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NM23-H2/NME2, Mouse anti-Human, Clone: CPTC-NME2-2, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79738-100UG
This item is not returnable.
View return policy
Description
NM23-H2/NME2 Monoclonal specifically detects NM23-H2/NME2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| NM23-H2/NME2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1-2 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| P22392, P22392 | |
| NME2 | |
| Recombinant full-length human NM23-H2/NME2 protein | |
| 100 μg | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| 4831 | |
| Store at 4°C. | |
| IgG2a κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| CPTC-NME2-2 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| EC 2.7.13.3, Histidine protein kinase NDKB, MGC2212, NDK B, NDKB, NDPKB, NDPK-B, nm23-H2, non-metastatic cells 2, protein (NM23B) expressed in, nucleoside diphosphate kinase B, protein (NM23) expressed in | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction