missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ NM23-H2/NME2 Antibody (NME2/6433), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals™ NBP3-26829-100ug

 View more versions of this product

Product Code. 30043382

  • £351.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

NM23-H2/NME2 Monoclonal antibody specifically detects NM23-H2/NME2 in Human samples. It is validated for Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NM23-H2/NME2
Monoclonal
0.2 mg/mL
Western Blot 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT
EC 2.7.13.3, Histidine protein kinase NDKB, MGC2212, NDK B, NDKB, NDPKB, NDPK-B, nm23-H2, non-metastatic cells 2, protein (NM23B) expressed in, nucleoside diphosphate kinase B, protein (NM23) expressed in
Recombinant full-length human NM23-H2/NME2 protein
100 μg
Primary
Human
Purified
Western Blot, Immunohistochemistry (Paraffin)
NME2/6433
Unconjugated
10 mM PBS with 0.05% BSA
Mouse
Protein A or G purified
RUO
4831
Store at 4°C.
IgG2 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.