missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NM23-H2/NME2 Antibody (4G7A8) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£175.00 - £435.00
Specifications
| Antigen | NM23-H2/NME2 |
|---|---|
| Clone | 4G7A8 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18246643
|
Novus Biologicals
NBP2-52519 |
0.1 mg |
£435.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18692829
|
Novus Biologicals
NBP2-52519-0.025mg |
0.025 mg |
£175.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NM23-H2/NME2 Monoclonal specifically detects NM23-H2/NME2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| NM23-H2/NME2 | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers | |
| PBS with 0.05% Sodium Azide | |
| 4831 | |
| Purified recombinant fragment of human NME2 (AA: FULL(1-152)) expressed in E. Coli. | |
| Primary | |
| 17.3 kDa |
| 4G7A8 | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| EC 2.7.13.3, Histidine protein kinase NDKB, MGC2212, NDK B, NDKB, NDPKB, NDPK-B, nm23-H2, non-metastatic cells 2, protein (NM23B) expressed in, nucleoside diphosphate kinase B, protein (NM23) expressed in | |
| NME2 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title