missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NM23-H1 Antibody (4B2), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
£161.00
Specifications
| Antigen | NM23-H1 |
|---|---|
| Clone | 4B2 |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
NM23-H1 Monoclonal specifically detects NM23-H1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| NM23-H1 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:200-1:1000, Immunohistochemistry-Paraffin 1:200-1:1000 | |
| Monoclonal | |
| Ascites | |
| RUO | |
| Human | |
| 4830 | |
| Purified recombinant fragment of human NME1 expressed in E. Coli. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 4B2 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Lipid and Metabolism | |
| EC 2.7.4.6, GAAD, Granzyme A-activated DNase, Metastasis inhibition factor nm23, NB, NBS, NDK A, NDKA, NDP kinase A, NDPKA, NDPK-A, NM23AWD, nm23-H1, non-metastatic cells 1, protein (NM23A) expressed in, nucleoside diphosphate kinase A, Tumor metastatic process-associated protein | |
| NME1 | |
| IgG1 | |
| Unpurified | |
| 17 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title