missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLRX1 Polyclonal Antibody

NLRX1 Polyclonal Antibody
Brand: Invitrogen PA5100025
This item is not returnable.
View return policy
Description
NLRX1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Specifications
| NLRX1 | |
| Polyclonal | |
| Unconjugated | |
| NLRX1 | |
| BC034204; Caterpiller protein 11.3; CLR11.3; DLNB26; FLJ21478; MGC131937; MGC21025; NLR family member X1; NLRX1; NOD26; NOD5; NOD9; NOD-like receptor X1; nucleotide-binding domain and leucine-rich repeat containing; nucleotide-binding oligomerization domain protein 26; Nucleotide-binding oligomerization domain protein 5; nucleotide-binding oligomerization domain protein 9; nucleotide-binding oligomerization domain, leucine rich repeat containing X1; OTTHUMP00000205768; OTTHUMP00000205769; RGD1311293 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 270151, 315599, 79671 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q3TL44, Q5FVQ8, Q86UT6 | |
| NLRX1 | |
| A synthesized peptide derived from human NLRX1(Accession Q86UT6), corresponding to amino acid residues R291-S341. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction