missing translation for 'onlineSavingsMsg'
Learn More

NLRP7 Antibody, DyLight 350, Novus Biologicals™

Product Code. 18425808 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425808 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18425808 Supplier Novus Biologicals Supplier No. NBP176290UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NLRP7 Polyclonal antibody specifically detects NLRP7 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen NLRP7
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 350
Formulation 50mM Sodium Borate
Gene Alias CLR19.4, MGC126471, NACHT, leucine rich repeat and PYD containing 7, NACHT, LRR and PYD containing protein 7, NACHT, LRR and PYD domains-containing protein 7, NALP7MGC126470, NLR family, pyrin domain containing 7, NOD12HYDM, Nucleotide-binding oligomerization domain protein 12, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 7, PAN7, PYPAF3FLJ94610, PYRIN-containing APAF1-like protein 3
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human NALP7. The immunogen is located within amino acids 90 - 140 of NALP7. Amino Acid Squence: LGDAEEDSELAKPGEKEG
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cell Biology, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 199713
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.