missing translation for 'onlineSavingsMsg'
Learn More

NLRP3/NALP3 Antibody (S07-9E9), Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Novus Biologicals NBP3-20025-100ul

 View more versions of this product

Product Code. 18684195

  • £500.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

NLRP3/NALP3 Monoclonal antibody specifically detects NLRP3/NALP3 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NLRP3/NALP3
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot
S07-9E9
Unconjugated
AII, AII/AVP, Angiotensin/vasopressin receptor AII/AVP-like, AVP, C1orf7, Caterpiller protein 1.1, CLR1.1, Cold autoinflammatory syndrome 1, Cold autoinflammatory syndrome 1 protein, Cryopyrin, Familial cold autoinflammatory syndrome, FCU, Leucine-rich repeat-, and PYD-containing protein 3, Muckle-Wells syndrome, MWS, NACHT, LRR and PYD containing protein 3, NACHT, LRR and PYD domains-containing protein 3, NALP3, NLR family, pyrin domain containing 3, NLRP3, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And Pyrin Domain Containing 3, PYPAF1, PYRIN-containing APAF1-like protein 1
Recombinant protein of human NLRP3/NALP3 (Uniprot # Q96P20)
100 μg
Alzheimers Research, Apoptosis, Cancer, Cardiovascular Biology, Cytokine Research, Immunology, Inflammation, Neurodegeneration, Signal Transduction
114548
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.