missing translation for 'onlineSavingsMsg'
Learn More

NLRP3/NALP3 Antibody, DyLight 488, Novus Biologicals™

Product Code. 18395530 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18395530 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18395530 Supplier Novus Biologicals Supplier No. NBP177080G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NLRP3/NALP3 Polyclonal specifically detects NLRP3/NALP3 in Human, Mouse, E. coli samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NLRP3/NALP3
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias AII, AII/AVP, Angiotensin/vasopressin receptor AII/AVP-like, AVP, C1orf7, Caterpiller protein 1.1, CLR1.1, Cold autoinflammatory syndrome 1, Cold autoinflammatory syndrome 1 protein, Cryopyrin, Familial cold autoinflammatory syndrome, FCU, Leucine-rich repeat-, and PYD-containing protein 3, Muckle-Wells syndrome, MWS, NACHT, LRR and PYD containing protein 3, NACHT, LRR and PYD domains-containing protein 3, NALP3, NLR family, pyrin domain containing 3, NLRP3, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And Pyrin Domain Containing 3, PYPAF1, PYRIN-containing APAF1-like protein 1
Gene Symbols NLRP3
Host Species Rabbit
Immunogen NLRP3/NALP3 Antibody was raised against a 16 amino acid synthetic peptide from near the amino terminus of human NALP3. The immunogen is located within amino acids 120 - 170 of NALP3.
Molecular Weight of Antigen 110 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Alzheimers Research, Apoptosis, Cancer, Cardiovascular Biology, Cytokine Research, Immunology, Inflammation, Neurodegeneration, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 114548
Target Species Human, Mouse, E. coli
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.