missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLRP3/NALP3 Antibody (CL0210) [Janelia Fluor™ 525], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52879JF525
This item is not returnable.
View return policy
Description
NLRP3/NALP3 Monoclonal antibody specifically detects NLRP3/NALP3 in Human samples. It is validated for Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| NLRP3/NALP3 | |
| Monoclonal | |
| Janelia Fluor 525 | |
| AII, AII/AVP, Angiotensin/vasopressin receptor AII/AVP-like, AVP, C1orf7, Caterpiller protein 1.1, CLR1.1, Cold autoinflammatory syndrome 1, Cold autoinflammatory syndrome 1 protein, Cryopyrin, Familial cold autoinflammatory syndrome, FCU, Leucine-rich repeat-, and PYD-containing protein 3, Muckle-Wells syndrome, MWS, NACHT, LRR and PYD containing protein 3, NACHT, LRR and PYD domains-containing protein 3, NALP3, NLR family, pyrin domain containing 3, NLRP3, Nucleotide-Binding Oligomerization Domain, Leucine Rich Repeat And Pyrin Domain Containing 3, PYPAF1, PYRIN-containing APAF1-like protein 1 | |
| This antibody was developed using a recombinant protein derived from Q96P20, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Alzheimers Research, Apoptosis, Cancer, Cardiovascular Biology, Cytokine Research, Immunology, Inflammation, Neurodegeneration, Signal Transduction | |
| 114548 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| CL0210 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction