missing translation for 'onlineSavingsMsg'
Learn More

NLRP13 Antibody, DyLight 680, Novus Biologicals™

Product Code. 18421825 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18421825 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18421825 Supplier Novus Biologicals Supplier No. NBP176294FR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NLRP13 Polyclonal antibody specifically detects NLRP13 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen NLRP13
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias CLR19.7, leucine rich repeat and pyrin domain containing 13, NACHT, leucine rich repeat and PYD containing 13, NACHT, LRR and PYD containing protein 13, NACHT, LRR and PYD domains-containing protein 13, NALP13, NLR family, pyrin domain containing 13, NOD14, nucleotide-binding oligomerization domain protein 14, PAN13
Host Species Rabbit
Immunogen Antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human NALP13. The immunogen is located within amino acids 150 - 200 of NALP13. Amino Acid Squence: QDPNQEEPEMLEEADHRR
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 126204
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.