missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLRP11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-69058-25ul
This item is not returnable.
View return policy
Description
NLRP11 Polyclonal antibody specifically detects NLRP11 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| NLRP11 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL | |
| CLR19.6, NACHT, leucine rich repeat and PYD containing 11, NALP11NACHT, LRR and PYD domains-containing protein 11, NLR family, pyrin domain containing 11, NOD17PYPAF7, Nucleotide-binding oligomerization domain protein 17, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 11, PAAD- and NACHT-containing protein 10, PAAD- and NACHT-containing protein 10B, PAAD-and NACHT domain-containing protein 10, PAN10FLJ26273, PYPAF6NACHT, LRR and PYD containing protein 11, PYRIN-containing APAF1-like protein 6 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: LRELHIFDNDLNGISERILSKALEHSSCKLRTLKLSYVSTASGFEDLLKALARNRSLTYLSINCTSISLNMFSLLHDILHEPTCQISHLSLMKCDLRASE | |
| 25 μL | |
| Immunology, Innate Immunity | |
| 204801 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction