missing translation for 'onlineSavingsMsg'
Learn More

NLRP10/Pynod/NALP10 Antibody, Novus Biologicals™

Product Code. p-200043629 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
25 μL
Unit Size:
0.1mL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18439101 25 μL 25µL
18206017 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18439101 Supplier Novus Biologicals Supplier No. NBP18555625ul

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody has been used in 1 publication

NLRP10/Pynod/NALP10 Polyclonal specifically detects NLRP10/Pynod/NALP10 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NLRP10/Pynod/NALP10
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias CLR11.1, NACHT, leucine rich repeat and PYD containing 10, NALP10Pynod, NLR family, pyrin domain containing 10, NOD8Nucleotide-binding oligomerization domain protein 8, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 10, PAN5, PYNODNACHT, LRR and PYD domains-containing protein 10
Gene Symbols NLRP10
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:RARYFSSYFTDEKQADRAFDIVQKNDILYKACQVPGICWVVCSWLQGQMERGKVVLETPRNSTDIFMAYVSTFLPPDDDGGCSELSRHRV
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 338322
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.