missing translation for 'onlineSavingsMsg'
Learn More

NLRP10/Pynod/NALP10 Antibody, Novus Biologicals™

Product Code. 18385861 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18385861 0.1 mg 0.10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18385861 Supplier Novus Biologicals Supplier No. NBP276809

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

NLRP10/Pynod/NALP10 Polyclonal specifically detects NLRP10/Pynod/NALP10 in Mouse samples. It is validated for Flow Cytometry, Peptide ELISA, Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NLRP10/Pynod/NALP10
Applications Flow Cytometry, ELISA, Immunofluorescence
Classification Polyclonal
Concentration 0.50 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 10 ug/ml, Peptide ELISA Detection Limit 1:128000, Immunofluorescence 10 ug/ml
Formulation Tris saline, pH7.3 with 0.5% bovine serum albumin. with 0.02% Sodium Azide
Gene Accession No. NP_780741.1
Gene Alias CLR11.1, NACHT, leucine rich repeat and PYD containing 10, NALP10Pynod, NLR family, pyrin domain containing 10, NOD8Nucleotide-binding oligomerization domain protein 8, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 10, PAN5, PYNODNACHT, LRR and PYD domains-containing protein 10
Gene Symbols NLRP10
Host Species Goat
Immunogen Peptide with sequence NDLEENSFKTLKFH-C, from the N Terminus of the protein sequence according to NP_780741.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Primary or Secondary Primary
Gene ID (Entrez) 338322.0
Target Species Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.