missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLRP1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12256-1-AP-150UL
This item is not returnable.
View return policy
Description
NALP1 is a member of the Ced-4 family of apoptosis proteins. Ced-family members contain a caspase recruitment domain (CARD) and are known to be key mediators of programmed cell death. The encoded protein contains a distinct N-terminal pyrin-like motif, which is possibly involved in protein-protein interactions. This protein interacts strongly with caspase 2 and weakly with caspase 9. Overexpression of this gene was demonstrated to induce apoptosis in cells. Multiple alternatively spliced transcript variants encoding distinct isoforms have been found for this gene, but the biological validity of some variants has not been determined.Specifications
| NLRP1 | |
| Polyclonal | |
| Unconjugated | |
| NLRP1 | |
| CARD7, CLR17.1, DEFCAP, DEFCAP L/S, KIAA0926, NAC, NALP1, NLRP1, PP1044, SLEV1, VAMAS1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22861 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9C000 | |
| NLRP1 | |
| NLRP1 Fusion Protein Ag2908 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction