missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLRP1/NALP1 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76656PCP
This item is not returnable.
View return policy
Description
NLRP1/NALP1 Polyclonal antibody specifically detects NLRP1/NALP1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Specifications
| NLRP1/NALP1 | |
| Polyclonal | |
| Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence | |
| CARD7SLEV1, Caspase recruitment domain-containing protein 7, CLR17.1, Death effector filament-forming ced-4-like apoptosis protein, DEFCAPVAMAS1, DKFZp586O1822, KIAA0926DEFCAP-L/S, NACHT, leucine rich repeat and PYD (pyrin domain) containing 1, NACHT, leucine rich repeat and PYD containing 1, NACHT, LRR and PYD containing protein 1, NACHT, LRR and PYD domains-containing protein 1, NACPP1044, NALP1caspase recruitment domain protein 7, NLR family, pyrin domain containing 1, Nucleotide-binding domain and caspase recruitment domain, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 1 | |
| Antibody was raised against a peptide corresponding to 13 amino acids near the carboxy terminus of human NALP1. The immunogen is located within the last 50 amino acids of NALP1. Amino Acid Squence: EKGSKKGLLPLSS | |
| 0.1 mL | |
| Apoptosis | |
| 22861 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunocytochemistry | |
| PerCP | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction