missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLRC5 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£360.00 - £553.00
Specifications
| Antigen | NLRC5 |
|---|---|
| Dilution | Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
| Applications | Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18302115
|
Novus Biologicals
NBP3-17901-25UL |
25 μg |
£360.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18397362
|
Novus Biologicals
NBP3-17901-100UL |
100 μg |
£553.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NLRC5 Polyclonal antibody specifically detects NLRC5 in Human samples. It is validated for ImmunofluorescenceSpecifications
| NLRC5 | |
| Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Immunology, Innate Immunity | |
| PBS, pH 7.2, 40% glycerol | |
| 84166 | |
| IgG | |
| Affinity purified |
| Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| Caterpiller Protein 16.1, CLR16.1, NLR Family CARD Domain Containing 5, NOD27, NOD4, NOD-Like Receptor C5, Nucleotide-Binding Oligomerization Domain Protein 27, Nucleotide-Binding Oligomerization Domain Protein 4, Nucleotide-Binding Oligomerization Domains 27, Protein NLRC5 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: GLVRCFSTLQWLFRLDISFESQHILLRGDKTSRDMWATGSLPDFPAAAKFLGFRQRCIPRSLCLSECPLEPPSLTRLCATLKDCPGPLELQLSCEFLSDQ | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title