missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NLRC5 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94762-0.1ml
This item is not returnable.
View return policy
Description
NLRC5 Polyclonal antibody specifically detects NLRC5 in Mouse samples. It is validated for Western Blot
Specifications
| NLRC5 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| Caterpiller Protein 16.1, CLR16.1, NLR Family CARD Domain Containing 5, NOD27, NOD4, NOD-Like Receptor C5, Nucleotide-Binding Oligomerization Domain Protein 27, Nucleotide-Binding Oligomerization Domain Protein 4, Nucleotide-Binding Oligomerization Domains 27, Protein NLRC5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-240 of human NLRC5 (NP_115582.4). MDPVGLQLGNKNLWSCLVRLLTKDPEWLNAKMKFFLPNTDLDSRNETLDPEQRVILQLNKLHVQGSDTWQSFIHCVCMQLEVPLDLEVLLLSTFGYDDGFTSQLGAEGKSQPESQLHHGLKRPHQSCGSSPRRKQCKKQQLELAKKYLQLLRTSAQQRYRSQIPGSGQPHAFHQVYVPPILRRATASLDTPEGAIMGDVKVEDGADVSISDLFNTRVNKGPRVTVLLGKAGMGKTTLAHR | |
| 0.1 mL | |
| Immunology, Innate Immunity | |
| 84166 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction