missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NLRC4 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA14264

Product Code. 11558800

  • £439.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

PA1-4264 detects CLAN protein from human samples. This antibody detects both A and B isoforms. PA1-4264 has been successfully used in Western blot procedures. By Western blot this antibody detects a ~116 kDa protein representing CLAN-A and a ~43 kDa protein representing CLAN-B in lung lysate. PA1-4264 immunizing peptide corresponds to N-terminal sequence M N F I K D N S R A L I Q R M G M.

NLRC4 encodes a member of the caspase recruitment domain-containing NLR family. Family members play essential roles in innate immune response to a wide range of pathogenic organisms, tissue damage and other cellular stresses. Mutations in this gene result in autoinflammation with infantile enterocolitis. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NLRC4
Polyclonal
Unconjugated
Nlrc4
9530011P19Rik; AIFEC; CARD, LRR, and NACHT-containing protein; Card12; caspase recruitment domain family, member 12; caspase recruitment domain-containing protein 12; CLAN; Clan protein; CLAN1; CLANA; CLANB; CLANC; CLAND; CLR2.1; FCAS4; ice protease-activating factor; ICE-protease activating factor; ipaf; NLR family CARD domain containing 4; NLR family CARD domain-containing protein 4; NLR family, CARD domain containing 4; NLRC4; NOD-like receptor C4; nucleotide-binding oligomerization domain, leucine rich repeat and CARD domain containing 4; UNQ6189/PRO20215
Rabbit
Antigen affinity chromatography
RUO
58484
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Liquid
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9NPP4
Nlrc4
Synthetic Peptide corresponding to the N-terminal sequence M N F I K D N S R A L I Q R M G M.
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.