missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NLN Polyclonal Antibody
GREENER_CHOICE

Product Code. 15724791 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. Quantity unitSize
15724791 100 μL 100µL
1 options
This item is not returnable. View return policy

Product Code. 15724791

Brand: Invitrogen™ PA556882

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LRMTLGREVM SPLQAMSSYT VAGRNVLRWD LSPEQIKTRT EELIVQTKQV YDAVGMLGIE EVTYENCL Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 81%.

This gene encodes a member of the metallopeptidase M3 protein family that cleaves neurotensin at the Pro10-Tyr11 bond, leading to the formation of neurotensin(1-10) and neurotensin(11-13). The encoded protein is likely involved in the termination of the neurotensinergic signal in the central nervous system and in the gastrointestinal tract.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NLN
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Nln
Gene Accession No. Q9BYT8
Gene Alias 4930472G13Rik; AGTBP; angiotensin binding protein; angiotensin-binding protein; C79345; endopeptidase 24.16; EP24.16; KIAA1226; MEP; microsomal endopeptidase; Mitochondrial oligopeptidase M; MOP; neurolysin; neurolysin (metallopeptidase M3 family); neurolysin, mitochondrial; Neurotensin endopeptidase; NLN
Gene Symbols Nln
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NLN. Recombinant protein control fragment (Product #RP-95832).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57486
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.