missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NLN Monoclonal Antibody (OTI1D6)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525842
This item is not returnable.
View return policy
Description
NLN Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
This gene encodes a member of the metallopeptidase M3 protein family that cleaves neurotensin at the Pro10-Tyr11 bond, leading to the formation of neurotensin(1-10) and neurotensin(11-13). The encoded protein is likely involved in the termination of the neurotensinergic signal in the central nervous system and in the gastrointestinal tract.
Specifications
| NLN | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P42676, Q91YP2, Q9BYT8 | |
| Nln | |
| Full length human recombinant protein of NLN produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat, Monkey | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| OTI1D6 | |
| Unconjugated | |
| Nln | |
| 4930472G13Rik; AGTBP; angiotensin binding protein; angiotensin-binding protein; C79345; endopeptidase 24.16; EP24.16; KIAA1226; MEP; microsomal endopeptidase; Mitochondrial oligopeptidase M; MOP; neurolysin; neurolysin (metallopeptidase M3 family); neurolysin, mitochondrial; Neurotensin endopeptidase; NLN | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 117041, 57486, 75805 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction