Learn More
NLGN3 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 29319-1-AP-20UL
 View more versions of this product
Product Code. 18078975
  • £135.00 / 20µL
Description
The NLGN3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human NLGN3. This antibody recognizes human, mouse, rat antigen. The NLGN3 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
NLGN3
600 μg/mL
Rabbit
Primary
Human, Mouse, Rat
150 kDa
IgG
Western Blot, Immunohistochemistry, ELISA
ASPGX1, AUTSX1, Gliotactin homolog, HNL3, KIAA1480, neuroligin 3, NL3, NLGN3
20 μL
54413
Store at -20°C. Stable for one year after shipment.