missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NLE1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA578509
This item is not returnable.
View return policy
Description
Positive Control: 293T, 293T nuclear extract Predicted Reactivity: Mouse (96%), Rat (96%), Xenopus laevis (81%), Rhesus Monkey (99%), Chimpanzee (99%), Bovine (94%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
NLE1 plays a role in regulating Notch activity, in regulating the expression of CDKN1A and several members of the Wnt pathway, probably via its effects on Notch activity. NLE1 is required during embryogenesis for inner mass cell survival.
Specifications
| NLE1 | |
| Polyclonal | |
| Unconjugated | |
| NLE1 | |
| AL022765; BC018399; FLJ10458; HUSSY-07; l11Jus1; l11Jus4; Nle; Nle1; Notchless gene homolog; notchless homolog 1; notchless homolog 1 (Drosophila); notchless protein homolog 1; novel WD40 repeat containing protein; RGD1308301 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54475 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.71 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q9NVX2 | |
| NLE1 | |
| Recombinant protein encompassing a sequence within the center region of human NLE1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction