missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nkx6.3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5104209
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total NKX6.3.
The NKX family of homeodomain proteins controls numerous developmental processes. Members of the NKX6 subfamily, including NKX6-3, are involved in development of the central nervous system (CNS), gastrointestinal tract, and pancreas.
Specifications
| Nkx6.3 | |
| Polyclonal | |
| Unconjugated | |
| NKX6-3 | |
| 9130417I07Rik; homeobox protein Nkx-6.3; NK6 homeobox 3; NK6 transcription factor related, locus 3; Nkx6.3; NKX6-3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 157848 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A6NJ46 | |
| NKX6-3 | |
| A synthesized peptide derived from human NKX6-3(Accession A6NJ46), corresponding to amino acid residues R169-G219. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction