missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NKX6-2 Polyclonal Antibody

Product Code. 15643285
Change view
Click to view available options
Quantity:
400 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15643285 400 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15643285 Supplier Invitrogen™ Supplier No. PA548988

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NKX6-2 Polyclonal Antibody for Western Blot

Transcription factor with repressor activity involved in the regulation of axon-glial interactions at myelin paranodes in oligodendrocytes. Binds to the consensus DNA sequence 5'-(A/T)TTAATGA-3'. In oligodendrocytes, binds to MBP and PLP1 promoter regions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKX6-2
Applications Western Blot
Classification Polyclonal
Concentration 0.45 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene NKX6-2
Gene Accession No. Q9C056
Gene Alias Drosophila NK transcription factor related, gene family 6, locus 2; glial and testis specific homeobox; glial and testis-specific homeobox protein; GTX; homeobox 6B; homeobox protein NK-6 homolog B; Homeobox protein Nkx-6.2; NK homeobox family 6, B; NK6 homeobox 2; NK6 transcription factor related, locus 2; Nkx6.2; NKX6-2; NKX6B
Gene Symbols NKX6-2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 17-43 amino acids from the N-terminal region of human NKX6-2.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84504
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.