missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nkx6.1 Recombinant Rabbit Monoclonal Antibody (K01_2Y67)

Product Code. 30829203
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30829203 100 μL
30827078 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30829203 Supplier Invitrogen™ Supplier No. MA563092

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Nkx6.1 Recombinant Monoclonal Antibody for IHC (P)

NKX6.1 is a bi-functional transcription factor that has effects in both the central nervous system and in the pancreas. It has both expression and repression effects on genes, leading to the proper formation of motoneurons, oligodendrocytes, and insulin-secreting beta cells. It is known to interact with Pax6 and Pdx-1, probably at or near the 60 amino acid homeobox DNA-binding domain. Due to its obvious relationship to beta cells and CNS, NKX6.1 is becoming a more interesting target for therapies in diabetes and certain brain diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nkx6.1
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone K01_2Y67
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol, 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene NKX6-1
Gene Accession No. P78426
Gene Alias Drosophila NK transcription factor related, gene family 6, locus 1; homeobox protein NK-6 homolog A; Homeobox protein Nkx-6.1; homeodomain transcription factor Nkx6.1; NK homeo box, family 6, member A; NK homeobox, family 6, A; NK6 homeobox 1; NK6 transcription factor homolog A; NK6 transcription factor related locus 1; NK6 transcription factor related, locus 1; Nkx6.1; Nkx61; NKX6-1; Nkx6a
Gene Symbols NKX6-1
Host Species Rabbit
Immunogen Recombinant protein of human NKX6.1.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4825
Target Species Human
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.