missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKX6.1 Antibody (NKX61/2561), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79872V
This item is not returnable.
View return policy
Description
NKX6.1 Monoclonal specifically detects NKX6.1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| NKX6.1 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NKX6-1 | |
| Recombinant full-length human NKX6.1 protein (Uniprot: P78426) | |
| 0.1 mL | |
| Pancreatic Stem Cell Markers, Stem Cell Signaling Pathway, Transcription Factors and Regulators | |
| Human | |
| IgG2c κ |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| NKX61/2561 | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Homeobox protein NK-6 homolog A, NK homeo box, family 6, member A, NK homeobox (Drosophila), family 6, A, NK homeobox, family 6, A, NK6 homeobox 1, NK6 transcription factor homolog A, NK6 transcription factor related, locus 1, NK6 transcription factor related, locus 1 (Drosophila), NKX6.1, NKX6Ahomeobox protein Nkx-6.1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction