missing translation for 'onlineSavingsMsg'
Learn More

NKX6.1 Antibody, HRP, Novus Biologicals™

Product Code. 18377433 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377433 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18377433 Supplier Novus Biologicals Supplier No. NBP299125H

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NKX6.1 Polyclonal specifically detects NKX6.1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKX6.1
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate HRP
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias Homeobox protein NK-6 homolog A, NK homeo box, family 6, member A, NK homeobox (Drosophila), family 6, A, NK homeobox, family 6, A, NK6 homeobox 1, NK6 transcription factor homolog A, NK6 transcription factor related, locus 1, NK6 transcription factor related, locus 1 (Drosophila), NKX6.1, NKX6Ahomeobox protein Nkx-6.1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of human NKX6.1.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Pancreatic Stem Cell Markers, Stem Cell Signaling Pathway, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 4825
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.