missing translation for 'onlineSavingsMsg'
Learn More

NKX3-2 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£149.00 - £366.00

Specifications

Antigen NKX3-2
Dilution Western Blot 1:100 - 1:500
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18613221
View Documents
Novus Biologicals
NBP2-93813-0.02ml
0.02 mL
£149.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18629220
View Documents
Novus Biologicals
NBP2-93813-0.1ml
0.1 mL
£366.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

NKX3-2 Polyclonal antibody specifically detects NKX3-2 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

NKX3-2
Western Blot
Unconjugated
Rabbit
Apoptosis, Cell Biology
PBS (pH 7.3), 50% glycerol
579
IgG
Affinity purified
Western Blot 1:100 - 1:500
Polyclonal
Purified
RUO
Human, Mouse, Rat
Bagpipe homeobox protein homolog 1, BAPX1bagpipe homeobox homolog 1 (Drosophila), Homeobox protein NK-3 homolog B, homeobox protein Nkx-3.2, MGC138171, NK3 homeobox 2, NKX3.2, NKX3BSMMD
Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human NKX3-2 (NP_001180.1). MAVRGANTLTSFSIQAILNKKEERGGLAAPEGRPAPGGTAASVAAAPAVCCWRLFGERDAGALGGAEDSLLASPAGTRTAAGRTAESPEGWDSDSALSEENESRRRCADARGASGAGLAGGSLSLGQPVCELAASKDLEEEAAGRSDSEMSASVSGDRSPRTEDDGVGPRGAHVSALCSG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.