missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKX3.1 Mouse anti-Human, Clone: NKX3.1/2836, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07435-100ug
This item is not returnable.
View return policy
Description
NKX3.1 Monoclonal specifically detects NKX3.1 in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| NKX3.1 | |
| Monoclonal | |
| 0.2 mg/mL | |
| ELISA, Immunohistochemistry-Paraffin 1 to 2 μg/mL, Protein Array | |
| BAPX2homeobox protein Nkx-3.1, Homeobox protein NK-3 homolog A, NK homeobox (Drosophila), family 3, A, NK3 homeobox 1, NK3 transcription factor homolog A, NK3 transcription factor related, locus 1, NK3 transcription factor related, locus 1 (Drosophila), NKX3.1NKX3, NKX3ANK homeobox, family 3, A | |
| Recombinant fragment (around aa 92-224) of human NKX3.1 protein (exact sequence is proprietary) | |
| 100 μg | |
| Cancer, Chromatin Research, Prostate Cancer, Transcription Factors and Regulators | |
| 4824 | |
| Store at 4°C. | |
| IgG1 κ |
| ELISA, Immunohistochemistry (Paraffin), Peptide Array | |
| NKX3.1/2836 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction