Learn More
Invitrogen™ Nkx3.1 Monoclonal Antibody (4H4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA515618
Description
MA5-15618 targets NKX3A in indirect ELISA, FACS, IHC, and WB applications and shows reactivity with Human samples. The MA5-15618 immunogen is purified recombinant fragment of human NKX3A expressed in E. Coli.. MA5-15618 detects NKX3A which has a predicted molecular weight of approximately 26.3kDa.
The homeodomain-containing transcription factor NKX3-1 is a putative prostate tumor suppressor that is expressed in a largely prostate-specific and androgen-regulated manner. Loss of NKX3-1 protein expression is a common finding in human prostate carcinomas and prostatic intraepithelial neoplasia.
Specifications
| Nkx3.1 | |
| Monoclonal | |
| Conc. Not Determined | |
| Ascites with 0.03% sodium azide | |
| Q99801 | |
| NKX3-1 | |
| Purified recombinant fragment of human NKX3A expressed in E. Coli. | |
| RUO | |
| 4824 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| 4H4 | |
| Unconjugated | |
| NKX3-1 | |
| bagpipe; BAPX2; Bax; Drosophila NK-3 transcription factor, locus 1; homeobox protein NK-3 homolog A; Homeobox protein Nkx-3.1; NK homeobox, family 3, A; NK3 homeobox 1; NK3 transcription factor homolog A; NK3 transcription factor related, locus 1; NK-3 transcription factor, locus 1; NK-3 transcription factor, locus 1 (Drosophila); NKX3; NKX3.1; Nkx-3.1; Nkx3-1; NKX3-1 antibody; NKX3A; prostate-specific ag | |
| Mouse | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.