missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nkx3.1 Monoclonal Antibody (3 9)
Description
Nkx3.1 Monoclonal Antibody for ELISA, IHC, Western Blot
The homeodomain-containing transcription factor NKX3-1 is a putative prostate tumor suppressor that is expressed in a largely prostate-specific and androgen-regulated manner. Loss of NKX3-1 protein expression is a common finding in human prostate carcinomas and prostatic intraepithelial neoplasia.
Specifications
Specifications
| Antigen | Nkx3.1 |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | 3 9 |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.4 |
| Gene | NKX3-1 |
| Gene Accession No. | P97436, Q99801 |
| Gene Alias | bagpipe; BAPX2; Bax; Drosophila NK-3 transcription factor, locus 1; homeobox protein NK-3 homolog A; Homeobox protein Nkx-3.1; NK homeobox, family 3, A; NK3 homeobox 1; NK3 transcription factor homolog A; NK3 transcription factor related, locus 1; NK-3 transcription factor, locus 1; NK-3 transcription factor, locus 1 (Drosophila); NKX3; NKX3.1; Nkx-3.1; Nkx3-1; NKX3-1 antibody; NKX3A; prostate-specific ag |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?