missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
NKX3.1 Monoclonal specifically detects NKX3.1 in Human samples. It is validated for ELISA, Immunohistochemistry-Paraffin, Protein Array.
Specifications
Specifications
| Antigen | NKX3.1 |
| Applications | ELISA, Immunohistochemistry (Paraffin), Peptide Array |
| Classification | Monoclonal |
| Clone | NKX3.1/3347 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | ELISA, Immunohistochemistry-Paraffin : 1-2 ug/mL, Protein Array |
| Formulation | 10mM PBS |
| Gene Alias | BAPX2homeobox protein Nkx-3.1, Homeobox protein NK-3 homolog A, NK homeobox (Drosophila), family 3, A, NK3 homeobox 1, NK3 transcription factor homolog A, NK3 transcription factor related, locus 1, NK3 transcription factor related, locus 1 (Drosophila), NKX3.1NKX3, NKX3ANK homeobox, family 3, A |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?