missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKX2.8 Mouse anti-Human, Clone: NKX28/2547, Novus Biologicals™
Mouse Monoclonal Antibody
£196.00
Specifications
| Antigen | NKX2.8 |
|---|---|
| Clone | NKX28/2547 |
| Concentration | 0.2 mg/ml |
| Dilution | Western Blot 0.5 ug-1 ug/ml |
| Applications | Western Blot |
Description
NKX2.8 Monoclonal specifically detects NKX2.8 in Human samples. It is validated for Western Blot.Specifications
| NKX2.8 | |
| 0.2 mg/ml | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Homeobox protein NK-2 homolog H, homeobox protein Nkx-2.8, NK2 homeobox 8, NK-2 homolog H (Drosophila), NK2 transcription factor related, locus 8, NK2 transcription factor related, locus 8 (Drosophila), NKX-2.8, NKX2.8NK-2 homolog 8, Nkx2-9, NKX2G, NKX2HNK-2 homolog H | |
| Human recombinant NKX2.8 protein fragment (around aa 10-123) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
| NKX28/2547 | |
| Western Blot 0.5 ug-1 ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 26257 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title