missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKX2.8 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-31642
This item is not returnable.
View return policy
Description
NKX2.8 Polyclonal specifically detects NKX2.8 in Human samples. It is validated for Western Blot.
Specifications
| NKX2.8 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| NKX2-8 | |
| Recombinant protein encompassing a sequence within the center region of human NKX2.8. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| 26257 | |
| Human | |
| IgG |
| Western Blot | |
| 1.0mg/mL | |
| Western Blot 1:500-1:3000 | |
| Homeobox protein NK-2 homolog H, homeobox protein Nkx-2.8, NK2 homeobox 8, NK-2 homolog H (Drosophila), NK2 transcription factor related, locus 8, NK2 transcription factor related, locus 8 (Drosophila), NKX-2.8, NKX2.8NK-2 homolog 8, Nkx2-9, NKX2G, NKX2HNK-2 homolog H | |
| Rabbit | |
| 26 kDa | |
| 100 μL | |
| Primary | |
| Tested in human. Predicted cross-reactivity based on sequence identity: Rat (80%). Not yet tested in any other species. | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction