missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Nkx2.6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550047
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total NKX2-6 protein.
Members of the NK-2 family of homeodomain proteins are key regulators of growth and development in several tissues, including brain, heart and pancreas.Nkx-2.5, also designated cardiac specific homeobox protein (Csx), is a homolog of the Drosophila tinman protein and is essential for normal cardiovascular development. Expression of Nkx-2.5 during cardiomyogenesis is required for cardiac septation, in which a single atrium and ventricle are separated into four chambers. Nkx-2.5 binds to DNA as a monomer, a homodimer or as a heterodimer with Nkx-2.3 or Nkx-2.6, which suggests that the specific protein-protein interactions of Nkx-2.5 are involved in its transcriptional regulatory function. Nkx-2.6, also a homolog of the Drosophila tinman protein, is expressed in the caudal pharyngeal pouches,the caudal heart progenitors, the sinus venosus, the outflow tract of the heart and in a short segment of the gut between stages E8.5 and E10.5 of embryogenesis. Expression of Nkx-2.6 overlaps with that of Nkx-2.5 in the pharynx and heart. However, Nkx-2.6 mutant mice are viable and fertile, which suggests that Nkx-2.6 plays a compensatory function to Nkx-2.5.
Specifications
| Nkx2.6 | |
| Polyclonal | |
| Unconjugated | |
| NKX2-6 | |
| AV044928; CSX2; CTHM; Drosophila NK2 transcription factor related, locus 6; homeobox protein NK2 homolog F; homeobox protein NK-2 homolog F; homeobox protein NKX2.6; homeobox protein Nkx-2.6; NK2 homeobox 6; NK2 transcription factor related, locus 6; Nkx2.6; Nkx-2.6; Nkx2-6; Nkx2f; NKX4-2; tinman; tinman paralog; Tix | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 137814 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A6NCS4 | |
| NKX2-6 | |
| Synthesized peptide derived from internal of human NKX26. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction