missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nkx2.5 Polyclonal Antibody

Product Code. 15997414
Change view
Click to view available options
Quantity:
200 μL
Unit Size:
400µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15997414 200 μL 400µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15997414 Supplier Invitrogen™ Supplier No. PA549431

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Nkx2.5 Polyclonal Antibody for Flow, ICC/IF, IHC (P), Western Blot

This gene encodes a homeobox-containing transcription factor. This transcription factor functions in heart formation and development. Mutations in this gene cause atrial septal defect with atrioventricular conduction defect, and also tetralogy of Fallot, which are both heart malformation diseases. Mutations in this gene can also cause congenital hypothyroidism non-goitrous type 5, a non-autoimmune condition. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Nkx2.5
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene Nkx2-5
Gene Accession No. P42582, P52952
Gene Alias Cardiac-specific homeobox; cardiac-specific homeobox 1; CHNG5; CSX; CSX1; Drosophila NK2 transcription factor related, locus 5; FLJ52202; FLJ97166; FLJ97195; FLJ97197; FLJ99536; HLHS2; Homeobox protein CSX; homeobox protein NK-2 homolog E; homeobox protein NKX 2-5; homeobox protein Nkx-2.5; NK2 homeobox 5; NK2 transcription factor related locus 5; NK2 transcription factor related, locus 5; NKX 2-5; NKX2.5; Nkx-2.5; Nkx-25; NKX2-5; NKX2E; NKX4-1; rNKx-2.5; tinman; tinman paralog; VSD3
Gene Symbols Nkx2-5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 98-133 amino acids from the Central region of mouse Nkx2-5.
Purification Method Protein A, Antigen affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1482, 18091
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.