missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NKX2.5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24675SS
This item is not returnable.
View return policy
Description
NKX2.5 Polyclonal specifically detects NKX2.5 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| NKX2.5 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| cardiac-specific homeo box, Cardiac-specific homeobox, cardiac-specific homeobox 1, CHNG5, CSX1, CSXFLJ97195, FLJ52202, FLJ99536, Homeobox protein CSX, Homeobox protein NK-2 homolog E, homeobox protein Nkx-2.5, NK2 transcription factor related, locus 5 (Drosophila), NKX2.5FLJ97166, NKX2EFLJ97197, NKX4-1, tinman paralog | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 1482 | |
| Human, Primate | |
| Purified |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Western Blot 1-2 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
| P52952 | |
| NKX2-5 | |
| A portion of amino acids 300-324 of human NKX2-5 was used as the immunogen. | |
| 0.025 mg | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, chimpanzee, cat, cow, horse, pig and rhesus monkey, 88% homologous in mouse, rat and opossum and 77% homologous in Xenopus and chicken. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction