missing translation for 'onlineSavingsMsg'
Learn More

NKX2.5 Antibody (259415) [DyLight 405], Novus Biologicals™

Product Code. 30731367 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30731367 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30731367 Supplier Novus Biologicals Supplier No. FAB24441E

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NKX2.5 Monoclonal antibody specifically detects NKX2.5 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen NKX2.5
Applications Western Blot
Classification Monoclonal
Clone 259415
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias cardiac-specific homeo box, Cardiac-specific homeobox, cardiac-specific homeobox 1, CHNG5, CSX1, CSXFLJ97195, FLJ52202, FLJ99536, Homeobox protein CSX, Homeobox protein NK-2 homolog E, homeobox protein Nkx-2.5, NK2 transcription factor related, locus 5 (Drosophila), NKX2.5FLJ97166, NKX2EFLJ97197, NKX4-1, tinman paralog
Host Species Mouse
Immunogen E. coli-derived recombinant human NKX2.5, Gln24-Arg137, Accession # P52952
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1482
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.